accessibility menu, dialog, popup

UserName

CH3OSiClOH3COH3C

CAS RN 5926-26-1

IUPAC Name: (chloromethyl)trimethoxysilane
Synonyms: (chloromethyl)trimethoxysilane
Molecular Weight (g/mol): 170.66
Molecular Formula: C4H11ClO3Si
InChi Key: FPOSCXQHGOVVPD-UHFFFAOYSA-N
SMILES: CO[Si](CCl)(OC)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results