missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl Pivalate 99.0+%, TCI America™

Catalog No. P101225G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P101225G 25 g
P1012100G 100 g
P1012500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P101225G Supplier TCI America Supplier No. P101225G

Chemical Identifiers

CAS 18997-19-8
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.60
MDL Number MFCD00000884
InChI Key GGRHYQCXXYLUTL-UHFFFAOYSA-N
Synonym chloromethyl pivalate, propanoic acid, 2,2-dimethyl-, chloromethyl ester, pivalic acid chloromethyl ester, pivaloyloxymethyl chloride, pivaloylmethyl chloride, chloromethyl trimethylacetate, 2,2-dimethylpropionic acid chloromethyl ester, chioromethyl pivalate, chloromethyl pivaloate, chloro-methyl pivalate
PubChem CID 87885
IUPAC Name chloromethyl 2,2-dimethylpropanoate
SMILES CC(C)(C)C(=O)OCCl

Specifications

Color Yellow
Boiling Point 147°C
Quantity 25 g
UN Number 2924
Formula Weight 150.60
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloromethyl Pivalate [Amino-Protecting Agent]