accessibility menu, dialog, popup

UserName

CH3CH3H3COOCl

CAS RN 18997-19-8

IUPAC Name: chloromethyl 2,2-dimethylpropanoate
Synonyms: chloromethyl 2,2-dimethylpropanoate
Molecular Weight (g/mol): 150.6
Molecular Formula: C6H11ClO2
InChi Key: GGRHYQCXXYLUTL-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)OCCl
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)