missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl phenyl sulfone, 99%

Catalog No. AC428740250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428740250 25 g Glass bottle
AC428740050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC428740250 Supplier Thermo Scientific Chemicals Supplier No. 428740250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7205-98-3 | C7H7ClO2S | 190.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7205-98-3
Molecular Formula C7H7ClO2S
Molecular Weight (g/mol) 190.65
MDL Number MFCD00007551
InChI Key NXAIQSVCXQZNRY-UHFFFAOYSA-N
Synonym chloromethyl phenyl sulfone, chloromethyl sulfonyl benzene, phenyl chloromethyl sulfone, chloromethylphenylsulfone, benzene, chloromethyl sulfonyl, chloromethylphenyl sulfone, 1-chloromethylsulfonyl benzene, chloromethyl sulphonyl benzene
PubChem CID 81625
IUPAC Name chloromethylsulfonylbenzene
SMILES C1=CC=C(C=C1)S(=O)(=O)CCl

Specifications

Melting Point 51°C to 53°C
Flash Point 113°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5SO2CH2Cl
Quantity 25 g
Formula Weight 190.65
Percent Purity 99%
Chemical Name or Material Chloromethyl phenyl sulfone
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 230-581-8

RUO – Research Use Only