missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl phenyl sulfide, 97%

Catalog No. p-4228525
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC202360050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC202360050 Supplier Thermo Scientific Chemicals Supplier No. 202360050

CAS: 7205-91-6 | C7H7ClS | 158.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7205-91-6
Molecular Formula C7H7ClS
Molecular Weight (g/mol) 158.64
MDL Number MFCD00000921
InChI Key LLSMWLJPWFSMCP-UHFFFAOYSA-N
Synonym chloromethyl phenyl sulfide, chloromethylthiobenzene, alpha-chlorothioanisole, phenyl chloromethyl sulfide, chloromethyl phenyl sulphide, phenylthio methyl chloride, chloromethyl thio benzene, sulfide, chloromethyl phenyl, chloromethyl phenyl sulfane, benzene, chloromethyl thio
PubChem CID 81623
IUPAC Name chloromethylsulfanylbenzene
SMILES ClCSC1=CC=CC=C1

Specifications

Color Colorless to Yellow
Density 1.2300g/mL
Boiling Point 66°C (0.3 hPa)
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.595
Linear Formula C6H5SCH2Cl
Quantity 5 g
Beilstein 06,III,1002
Fieser 09,107; 06,571; 07,357; 08,472; 09,451; 10,384; 11,503; 12,468; 13,34; 14,177; 16,98; 17,327
Specific Gravity 1.23
Formula Weight 158.65
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Chloromethyl phenyl sulfide
Show More Show Less
EINECSNumber 230-579-7

RUO – Research Use Only