missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloromethyl Isopropyl Carbonate 98.0+%, TCI America™
$100.67 - $301.98
Chemical Identifiers
| CAS | 35180-01-9 |
|---|---|
| Molecular Formula | C5H9ClO3 |
| Molecular Weight (g/mol) | 152.57 |
| MDL Number | MFCD07375443 |
| InChI Key | JHYNXXBAHWPABC-UHFFFAOYSA-N |
| Synonym | Carbonic Acid Chloromethyl Isopropyl Ester |
| PubChem CID | 14787051 |
| IUPAC Name | chloromethyl propan-2-yl carbonate |
| SMILES | CC(C)OC(=O)OCCl |
Chemical Identifiers
| 35180-01-9 | |
| 152.57 | |
| JHYNXXBAHWPABC-UHFFFAOYSA-N | |
| 14787051 | |
| CC(C)OC(=O)OCCl |
| C5H9ClO3 | |
| MFCD07375443 | |
| Carbonic Acid Chloromethyl Isopropyl Ester | |
| chloromethyl propan-2-yl carbonate |
Specifications
| 35180-01-9 | |
| C5H9ClO3 | |
| 25 g | |
| Carbonic Acid Chloromethyl Isopropyl Ester | |
| CC(C)OC(=O)OCCl | |
| 152.57 | |
| 152.57 | |
| Liquid |
| Yellow | |
| MFCD07375443 | |
| 2924 | |
| JHYNXXBAHWPABC-UHFFFAOYSA-N | |
| chloromethyl propan-2-yl carbonate | |
| 14787051 | |
| ≥98.0% (GC) | |
| Chloromethyl Isopropyl Carbonate |
Safety and Handling
TSCA : No