accessibility menu, dialog, popup

UserName

CAS RN 35180-01-9

IUPAC Name: chloromethyl propan-2-yl carbonate
Synonyms: chloromethyl isopropyl carbonate
Molecular Weight (g/mol): 152.57
Molecular Formula: C5H9ClO3
InChi Key: JHYNXXBAHWPABC-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)OCCl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results