missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl Isopropyl Carbonate 98.0+%, TCI America™

Catalog No. C262625G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C262625G 25 g
C2626100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C262625G Supplier TCI America Supplier No. C262625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 35180-01-9
Molecular Formula C5H9ClO3
Molecular Weight (g/mol) 152.57
MDL Number MFCD07375443
InChI Key JHYNXXBAHWPABC-UHFFFAOYSA-N
Synonym Carbonic Acid Chloromethyl Isopropyl Ester
PubChem CID 14787051
IUPAC Name chloromethyl propan-2-yl carbonate
SMILES CC(C)OC(=O)OCCl

Specifications

Color Yellow
Quantity 25 g
UN Number 2924
Formula Weight 152.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloromethyl Isopropyl Carbonate