missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloromethyl Chlorosulfonate 98.0+%, TCI America™
Supplier: TCI America C26605G
Specifications
| Chloromethyl Chlorosulfonate | |
| Yellow | |
| CH2Cl2O3S | |
| 5 g | |
| chloromethyl chlorosulfate, chloromethyl chlorosulphate, chloromethylchlorosulfate, chloromethyl chlorosulfonate, chloromethyl chloranesulfonate, unii-n615783bnf, chloromethyl sulfochloridate, chlorosulfuric acid, chloromethyl ester, chloro chlorosulfonyloxy methane, chlorosulfuric acid chloromethyl ester | |
| C(OS(=O)(=O)Cl)Cl | |
| 164.984 | |
| 164.98 | |
| Liquid |
| 49715-04-0 | |
| 50°C | |
| MFCD00041547 | |
| 3265 | |
| PJBIHXWYDMFGCV-UHFFFAOYSA-N | |
| chloro(chlorosulfonyloxy)methane | |
| 2733556 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 49715-04-0 | |
| 164.984 | |
| PJBIHXWYDMFGCV-UHFFFAOYSA-N | |
| 2733556 | |
| C(OS(=O)(=O)Cl)Cl |
| CH2Cl2O3S | |
| MFCD00041547 | |
| chloromethyl chlorosulfate, chloromethyl chlorosulphate, chloromethylchlorosulfate, chloromethyl chlorosulfonate, chloromethyl chloranesulfonate, unii-n615783bnf, chloromethyl sulfochloridate, chlorosulfuric acid, chloromethyl ester, chloro chlorosulfonyloxy methane, chlorosulfuric acid chloromethyl ester | |
| chloro(chlorosulfonyloxy)methane |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator