missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl Chlorosulfonate 98.0+%, TCI America™

Catalog No. p-7125584
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26605G 5 g
C266025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C26605G Supplier TCI America Supplier No. C26605G
Only null left

Chemical Identifiers

CAS 49715-04-0
Molecular Formula CH2Cl2O3S
Molecular Weight (g/mol) 164.984
MDL Number MFCD00041547
InChI Key PJBIHXWYDMFGCV-UHFFFAOYSA-N
Synonym chloromethyl chlorosulfate, chloromethyl chlorosulphate, chloromethylchlorosulfate, chloromethyl chlorosulfonate, chloromethyl chloranesulfonate, unii-n615783bnf, chloromethyl sulfochloridate, chlorosulfuric acid, chloromethyl ester, chloro chlorosulfonyloxy methane, chlorosulfuric acid chloromethyl ester
PubChem CID 2733556
IUPAC Name chloro(chlorosulfonyloxy)methane
SMILES C(OS(=O)(=O)Cl)Cl

Specifications

Color Yellow
Boiling Point 50°C
Quantity 5 g
UN Number 3265
Formula Weight 164.98
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Chloromethyl Chlorosulfonate
TSCA No
Recommended Storage Refrigerator