missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl Chloroformate 95.0+%, TCI America™

Catalog No. p-7124952
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C139625G 25 g
C1396100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C139625G Supplier TCI America Supplier No. C139625G
Only null left

Chemical Identifiers

CAS 22128-62-7
Molecular Formula C2H2Cl2O2
Molecular Weight (g/mol) 128.936
MDL Number MFCD00077688
InChI Key JYWJULGYGOLCGW-UHFFFAOYSA-N
Synonym chloromethyl chloroformate, chloromethylchloroformate, chloroformic acid chloromethyl ester, carbonochloridic acid, chloromethyl ester, palite, chlormethylchlorformiat, clco2ch2cl, chlormethylchlorocarbonat, chloromethyl choroformate, choromethyl chloroformate
PubChem CID 62754
IUPAC Name chloromethyl carbonochloridate
SMILES C(OC(=O)Cl)Cl

Specifications

Color Yellow
Boiling Point 108°C
Quantity 25 g
UN Number 2745
Formula Weight 128.94
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Chloromethyl Chloroformate
TSCA No
Recommended Storage Refrigerator