accessibility menu, dialog, popup

UserName

ClOClO

CAS RN 22128-62-7

IUPAC Name: chloromethyl carbonochloridate
Synonyms: chloromethyl chloroformate
Molecular Weight (g/mol): 128.94
Molecular Formula: C2H2Cl2O2
InChi Key: JYWJULGYGOLCGW-UHFFFAOYSA-N
SMILES: ClCOC(Cl)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)