missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloromethyl chloroformate, 97%
Description
Chloromethyl chloroformate is used as an intermediate in pharmaceuticals and in manufacture other chemicals. It acts as a reactant in the synthesis of aminocarbonyloxymethyl esters of diclofenac, flufenamic acid and in monomethoxypoly(ethyleneglycol) prodrugs of cyclosporin A.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chloromethyl chloroformate is used as an intermediate in pharmaceuticals and in manufacture other chemicals. It acts as a reactant in the synthesis of aminocarbonyloxymethyl esters of diclofenac, flufenamic acid and in monomethoxypoly(ethyleneglycol) prodrugs of cyclosporin A.
Solubility
Slightly miscible with water.
Notes
Incompatible with oxidizing agents and water.
Chloromethyl chloroformate is used as an intermediate in pharmaceuticals and in manufacture other chemicals. It acts as a reactant in the synthesis of aminocarbonyloxymethyl esters of diclofenac, flufenamic acid and in monomethoxypoly(ethyleneglycol) prodrugs of cyclosporin A.
Solubility
Slightly miscible with water.
Notes
Incompatible with oxidizing agents and water.
Specifications
Specifications
| Melting Point | <-20°C |
| Density | 1.465 |
| Boiling Point | 107°C to 108°C |
| Flash Point | 95°C (203°F) |
| Odor | Acrid |
| Refractive Index | 1.4295 |
| Linear Formula | ClCO2CH2Cl |
| Quantity | 10 g |
| UN Number | UN2745 |
| Beilstein | 506426 |
| Show More |
Safety and Handling
| Hazard Category | H302-H314-H330-H335 |
| Hazard Statement | GHS H Statement H331-H314-H318-H302 Toxic if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROMETHYL CHLOROFORMATE |
| EINECSNumber | 244-793-3 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only