missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl chloroformate, 97%

Catalog No. AAL0952709
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0952709 10 g
AAL0952718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0952709 Supplier Thermo Scientific Chemicals Supplier No. L0952709
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 22128-62-7 | C2H2Cl2O2 | 128.936 g/mol

Chloromethyl chloroformate is used as an intermediate in pharmaceuticals and in manufacture other chemicals. It acts as a reactant in the synthesis of aminocarbonyloxymethyl esters of diclofenac, flufenamic acid and in monomethoxypoly(ethyleneglycol) prodrugs of cyclosporin A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloromethyl chloroformate is used as an intermediate in pharmaceuticals and in manufacture other chemicals. It acts as a reactant in the synthesis of aminocarbonyloxymethyl esters of diclofenac, flufenamic acid and in monomethoxypoly(ethyleneglycol) prodrugs of cyclosporin A.

Solubility
Slightly miscible with water.

Notes
Incompatible with oxidizing agents and water.

Chemical Identifiers

CAS 22128-62-7
Molecular Formula C2H2Cl2O2
Molecular Weight (g/mol) 128.936
MDL Number MFCD00077688
InChI Key JYWJULGYGOLCGW-UHFFFAOYSA-N
Synonym chloromethyl chloroformate, chloromethylchloroformate, chloroformic acid chloromethyl ester, carbonochloridic acid, chloromethyl ester, palite, chlormethylchlorformiat, clco2ch2cl, chlormethylchlorocarbonat, chloromethyl choroformate, choromethyl chloroformate
PubChem CID 62754
IUPAC Name chloromethyl carbonochloridate
SMILES C(OC(=O)Cl)Cl

Specifications

Melting Point <-20°C
Density 1.465
Boiling Point 107°C to 108°C
Flash Point 95°C (203°F)
Odor Acrid
Refractive Index 1.4295
Linear Formula ClCO2CH2Cl
Quantity 10 g
UN Number UN2745
Beilstein 506426
Sensitivity Moisture sensitive
Solubility Information Slightly miscible with water.
Formula Weight 128.94
Percent Purity 97%
Chemical Name or Material Chloromethyl chloroformate
Show More Show Less
Hazard Category H302-H314-H330-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROMETHYL CHLOROFORMATE
EINECSNumber 244-793-3
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only