missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl Butyrate 98.0+%, TCI America™

Catalog No. C284125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C284125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C284125G Supplier TCI America Supplier No. C284125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33657-49-7
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.575
MDL Number MFCD00216961
InChI Key BDPZFQLKFUONAG-UHFFFAOYSA-N
Synonym chloromethyl butyrate, chloromethylbutyrate, butyric acid chloromethyl ester, butanoic acid, chloromethyl ester, chloromethyl n-butanoate, butyryloxymethyl chloride, acmc-1agry, ksc492g7j, butanoic acid,chloromethyl ester, n-butanoic acid chloromethyl ester
PubChem CID 2725008
IUPAC Name chloromethyl butanoate
SMILES CCCC(=O)OCCl

Specifications

Color Yellow
Boiling Point 149°C
Quantity 25 g
UN Number 2924
Formula Weight 136.58
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloromethyl Butyrate