accessibility menu, dialog, popup

UserName

CAS RN 33657-49-7

IUPAC Name: chloromethyl butanoate
Synonyms: chloromethyl butanoate
Molecular Weight (g/mol): 136.58
Molecular Formula: C5H9ClO2
InChi Key: BDPZFQLKFUONAG-UHFFFAOYSA-N
SMILES: CCCC(=O)OCCl
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)