missing translation for 'onlineSavingsMsg'
Learn More

Chloromethyl butyrate, 99%

Catalog No. AC295580050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC295580050 5 g Glass bottle
AC295580250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC295580050 Supplier Thermo Scientific Chemicals Supplier No. 295580050

CAS: 33657-49-7 | C5H9ClO2 | 136.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 33657-49-7
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.58
MDL Number MFCD00216961
InChI Key BDPZFQLKFUONAG-UHFFFAOYSA-N
Synonym chloromethyl butyrate, chloromethylbutyrate, butyric acid chloromethyl ester, butanoic acid, chloromethyl ester, chloromethyl n-butanoate, butyryloxymethyl chloride, acmc-1agry, ksc492g7j, butanoic acid,chloromethyl ester, n-butanoic acid chloromethyl ester
PubChem CID 2725008
IUPAC Name chloromethyl butanoate
SMILES CCCC(=O)OCCl

Specifications

Color Colorless
Density 1.0740g/mL
Boiling Point 150.0°C (993.0 mbar)
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4190 to 1.4210
Linear Formula CH3(CH2)2C(O)OCH2Cl
Quantity 5 g
Specific Gravity 1.074
Solubility Information Solubility in water: 3g/L (20°C)
Formula Weight 136.58
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Chloromethyl butyrate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only