missing translation for 'onlineSavingsMsg'
Learn More

Chlorohydroquinone 85.0+%, TCI America™

Catalog No. C018525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C018525G 25 g
C0185500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C018525G Supplier TCI America Supplier No. C018525G

Chemical Identifiers

CAS 615-67-8
Molecular Formula C6H5ClO2
Molecular Weight (g/mol) 144.554
MDL Number MFCD00002341
InChI Key AJPXTSMULZANCB-UHFFFAOYSA-N
Synonym chlorohydroquinone, 2-chlorohydroquinone, monochlorohydroquinone, chloroquinol, 1,4-benzenediol, 2-chloro, 2-chloro-1,4-benzenediol, hydroquinone, chloro, 2-chloro-1,4-dihydroxybenzene, 2,5-dihydroxychlorobenzene, 1,4-dihydroxy-2-chlorobenzene
PubChem CID 301
ChEBI CHEBI:27675
IUPAC Name 2-chlorobenzene-1,4-diol
SMILES C1=CC(=C(C=C1O)Cl)O

Specifications

Melting Point 105°C
Boiling Point 263°C
Quantity 25 g
Formula Weight 144.55
Percent Purity ≥85.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Chlorohydroquinone
EINECSNumber (3)-2847
RTECSNumber MX4800000
TSCA Yes