missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroformamidine Hydrochloride 98.0+%, TCI America™
$69.31 - $204.07
Chemical Identifiers
| CAS | 29671-92-9 |
|---|---|
| Molecular Formula | CH4Cl2N2 |
| Molecular Weight (g/mol) | 114.957 |
| MDL Number | MFCD00035527 |
| InChI Key | FUQFHOLPJJETAP-UHFFFAOYSA-N |
| PubChem CID | 3084509 |
| IUPAC Name | carbamimidoyl chloride;hydrochloride |
| SMILES | C(=N)(N)Cl.Cl |
Chemical Identifiers
| 29671-92-9 | |
| 114.957 | |
| FUQFHOLPJJETAP-UHFFFAOYSA-N | |
| carbamimidoyl chloride;hydrochloride |
| CH4Cl2N2 | |
| MFCD00035527 | |
| 3084509 | |
| C(=N)(N)Cl.Cl |
Specifications
| 29671-92-9 | |
| White-Yellow | |
| MFCD00035527 | |
| FUQFHOLPJJETAP-UHFFFAOYSA-N | |
| carbamimidoyl chloride;hydrochloride | |
| 3084509 | |
| ≥98.0% (N,T) | |
| Chloroformamidine Hydrochloride |
| 180°C | |
| CH4Cl2N2 | |
| 5 g | |
| C(=N)(N)Cl.Cl | |
| 114.957 | |
| 114.96 | |
| Crystalline Powder |
Safety and Handling
TSCA : No