missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroformamidine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America C319925G
Specifications
| Chloroformamidine Hydrochloride | |
| 180°C | |
| CH4Cl2N2 | |
| 25 g | |
| C(=N)(N)Cl.Cl | |
| 114.957 | |
| 114.96 | |
| Crystalline Powder |
| 29671-92-9 | |
| White-Yellow | |
| MFCD00035527 | |
| FUQFHOLPJJETAP-UHFFFAOYSA-N | |
| carbamimidoyl chloride;hydrochloride | |
| 3084509 | |
| ≥98.0% (N,T) |
Chemical Identifiers
| 29671-92-9 | |
| 114.957 | |
| FUQFHOLPJJETAP-UHFFFAOYSA-N | |
| carbamimidoyl chloride;hydrochloride |
| CH4Cl2N2 | |
| MFCD00035527 | |
| 3084509 | |
| C(=N)(N)Cl.Cl |
Safety and Handling
TSCA : No