missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform Solution, NMR Reference Standard, 5% in Acetone-d6 (99.9 atom% D), MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| Packaging | 5 mm O.D. tube |
| CAS | 666-52-4 |
| Flash Point | -17°C |
| MDL Number | MFCD00000826 |
| Type | NMR Reference Standard |
| Quantity | 0.7 mL |
| Grade | NMR Reference Standard |
| Concentration or Composition (by Analyte or Components) | 5% in acetone-d 6 (99.9 atom % D) |