missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, >=99.8%, ACS reagent, stab. with EtOH, meets the requir. of Reag.Ph.Eur.

Catalog No. p-217334
Change view
Click to view available options
Quantity:
25 mL
500 mL
1 L
2.5 L
4 L
Packaging:
Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC423550250 25 mL Glass Bottle
AC423555000 500 mL Glass Bottle
AC423550010 1 L Glass Bottle
AC423550025 2.5 L Glass Bottle
AC423550040 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AC423550250 Supplier Thermo Scientific Chemicals Supplier No. 423550250
Only null left

CAS: 67-66-3 | CHCl3 | 119.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 67-66-3 , 64-17-5
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
SMILES ClC(Cl)Cl

Specifications

Melting Point -63.0°C
Decomposition Information 290°C
Density 1.4800g/mL
Boiling Point 61.0°C
Quantity 25 mL
Infrared Spectrum Conforms to Ref Report
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4443 to 1.4463
Assay Percent Range 99.8+%
Residue after Evaporation ≤0.001%
Specific Gravity 1.48
Linear Formula CHCl3
Vapor Pressure Vapor Pressure: 213mbar at 20°C
Viscosity 0.56 mPa.s (20°C)
Beilstein 01, 61
Fieser 01,130; 08,92; 15,84
Merck Index 15, 2142
Solubility Information Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in aceton
IUPAC Name chloroform
Formula Weight 119.38
Percent Purity ≥99.8%
Grade ACS Reagent
Chemical Name or Material Chloroform, Stabilized
Show More Show Less
Hazard Category Warning
Hazard Statement GHS P Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.
Precautionary Statement GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 200-663-8
RTECSNumber FS9100000
TSCA TSCA

RUO – Research Use Only