Learn More
Chloroform, >=99.8%, ACS reagent, stab. with EtOH, meets the requir. of Reag.Ph.Eur.
CAS: 67-66-3 | CHCl3 | 119.37 g/mol
Supplier: Thermo Scientific Chemicals 423550040
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Chloroform | |
| Stabilized | |
| 290°C | |
| 61.0°C | |
| Conforms to Ref Report | |
| Liquid | |
| ≤0.001% | |
| 1.48 | |
| CHCl3 | |
| Vapor Pressure: 213mbar at 20°C | |
| 01, 61 | |
| 15, 2142 | |
| Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in aceton | |
| ClC(Cl)Cl | |
| 119.37 | |
| CHEBI:35255 | |
| ≥99.8% |
| -63.0°C | |
| 67-66-3,64-17-5 | |
| 1.4800g/mL | |
| 4 L | |
| Glass Bottle | |
| 1.4443 to 1.4463 | |
| 99.8+% | |
| CHCl3 | |
| MFCD00000826 | |
| 0.56 mPa.s (20°C) | |
| 01,130; 08,92; 15,84 | |
| trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
| HEDRZPFGACZZDS-UHFFFAOYSA-N | |
| trichloromethane | |
| 6212 | |
| 119.38 | |
| ACS Reagent |
Chemical Identifiers
| 67-66-3 | |
| 119.37 | |
| HEDRZPFGACZZDS-UHFFFAOYSA-N | |
| 6212 | |
| ClC(Cl)Cl |
| CHCl3 | |
| MFCD00000826 | |
| trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
| CHEBI:35255 |
Safety and Handling
GHS P Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
Warning
EINECSNumber : 200-663-8
RTECSNumber : FS9100000
TSCA : TSCA
RUO – Research Use Only