missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, >=99.8%, ACS reagent, stab. with EtOH, meets the requir. of Reag.Ph.Eur.

CAS: 67-66-3 | CHCl3 | 119.37 g/mol

Supplier:  Thermo Scientific Chemicals 423550010

 Afficher plus de versions de ce produit

Numéro de catalogue. AC423550010

Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

Chloroform
Stabilized
290°C
61.0°C
Conforms to Ref Report
Liquid
≤0.001%
1.48
CHCl3
Vapor Pressure: 213mbar at 20°C
01, 61
15, 2142
Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in aceton
ClC(Cl)Cl
119.37
CHEBI:35255
≥99.8%
-63.0°C
67-66-3,64-17-5
1.4800g/mL
1 L
Glass Bottle
1.4443 to 1.4463
99.8+%
CHCl3
MFCD00000826
0.56 mPa.s (20°C)
01,130; 08,92; 15,84
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
HEDRZPFGACZZDS-UHFFFAOYSA-N
chloroform
6212
119.38
ACS Reagent

Identifiants chimiques

67-66-3
119.37
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
ClC(Cl)Cl
CHCl3
MFCD00000826
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
CHEBI:35255
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS P Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.

GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

Warning

missing translation for 'einecsNumber' : 200-663-8

missing translation for 'rtecsNumber' : FS9100000

missing translation for 'tsca' : TSCA

FDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only