missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, for HPLC, stabilized with amylene

Catalog No. AC610030040
Change view
Click to view available options
Quantity:
4 L
Packaging:
EcoSafPak™
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC610030040 4 L EcoSafPak™
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC610030040 Supplier Thermo Scientific Chemicals Supplier No. 610030040

missing translation for 'validMainframeMsg'

CAS: 67-66-3 | CHCl3 | 119.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC

Chemical Identifiers

CAS 67-66-3 , 513-35-9
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Melting Point -63°C
Absorbance (1 cm cell vs water), 0.01 max. at 280nm, 0.15 max. at 254nm, 0.32 max. at 250nm, 1 max. at 245nm
CAS Min % 99.0, 1.0
Decomposition Information 290°C
Density 1.4800g/mL
Quantity 4 L
Identification Pass Test
Packaging EcoSafPak™
Refractive Index 1.4430 - 1.4460
Residue after Evaporation 2ppm max.
Linear Formula CHCl3
Vapor Pressure Vapor Pressure: 213mbar at 20°C
Water 0.02% max.
Beilstein 01, 61
Fieser 01,130; 08,92; 15,84
Merck Index 15, 2142
Boiling Point 61°C
Formula Weight 119.38
Percent Purity ≥99.8%
Grade HPLC
Chemical Name or Material Chloroform, 99.8%
Show More Show Less
Hazard Category Signal Word: WARNING!
Hazard Statement GHS H Statement:
Causes skin irritation.
Suspected of causing cancer.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement Emergency Overview
Harmful if swallowed. Irritating to eyes, respiratory system and skin.
NFPA
Health: 2
Flammability:

RUO – Research Use Only