missing translation for 'onlineSavingsMsg'
Learn More

Chloroform (HPLC), Fisher Chemical™

Catalog No. C607SK4
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Safe-Cote™ Amber Glass 
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
C607SK4 4 L Safe-Cote™ Amber Glass 
C6071 1 L Glass Bottle
C6074 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C607SK4 Supplier Thermo Fisher Scientific Supplier No. BPC607SK4
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

HPLC mobile phase for analytical and preparative separations

Chemical Identifiers

CAS 67-66-3 , 513-35-9
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Melting Point -63°C
Absorbance 0.01 AU max., 0.01 AU max. at 280nm, 0.05 AU max. at 270nm, 0.15 AU max. at 254nm, 0.15 AU max. at 260nm, 0.25 AU max. at 255nm, 0.32 AU max. at 250nm, 1.00 AU max. at 245nm, Pass Test
Color Colorless
Density 1.480g/cm³
Quantity 4 L
Identification Pass Test
Packaging Safe-Cote™ Amber Glass 
Physical Form Liquid
Refractive Index 1.4430 to 1.4460 (at 25°C)
Residue after Evaporation 2ppm max.
Assay Percent Range 99.8% min by GC
Vapor Pressure 213mbar at 20°C
Viscosity 0.56 mPa/s at 20°C
Water 0.02% max.
Boiling Point 61°C
Formula Weight 119.38
Percent Purity ≥99.8%
Grade Mobile phase for HPLC applications
Concentration or Composition (by Analyte or Components) Approx. 50ppm Amylene as Preservative
Chemical Name or Material Chloroform, Approx. 50ppm Pentene as Preservative
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Precautionary Statement Emergency Overview
Harmful if swallowed. Irritating to eyes, respiratory system and skin. Symptoms of overexposure are dizziness, headache, tiredness, nausea, unconsciousness, cessation of breathing.
<special id
DOTInformation DOT Class 6.1, : Poison