missing translation for 'onlineSavingsMsg'
Learn More

Chloroform (Approx. 0.75% Ethanol as Preservative/HPLC), Fisher Chemical™

Catalog No. p-215822
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Safe-Cote™ Amber Glass 
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
C6061 1 L Glass Bottle
C6064 4 L Glass Bottle
C606SK4 4 L Safe-Cote™ Amber Glass 
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C6061 Supplier Thermo Fisher Scientific Supplier No. FLC6061
Only null left

HPLC mobile phase for analytical and preparative separations

Chemical Identifiers

CAS 67-66-3 , 64-17-5
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Melting Point -63°C
Absorbance 0.13 AU max. at 254nm, 0.25 AU max. at 255nm, 0.15 AU max. at 260nm, 0.05 AU max. at 270nm, 0.01 AU max. (290 to 400nm), Pass Test, 1.00 AU max. at 244nm, 0.80 AU max. at 245nm, 0.28 AU max. at 250nm
Color Colorless
Decomposition Information 290°C
Density 1.480g/cm³
Quantity 1 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4420 to 1.4450 (at 25°C)
Residue after Evaporation 2ppm max.
Assay Percent Range 99.8% min by GC
Vapor Pressure 213mbar at 20°C
Viscosity 0.56 mPa/s at 20°C
UN Number 1888
Water 0.02% max.
Boiling Point 61°C
Formula Weight 119.38
Percent Purity ≥99.8%
Grade Mobile phase for HPLC applications
Concentration or Composition (by Analyte or Components) Approx. 0.75% Ethanol as Preservative
Chemical Name or Material Chloroform, Approx. 0.75% Ethanol as Preservative
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. Irritating to eyes and skin. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. May be harmful if inhaled. May cause irritation of respiratory tract. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.

NFPA

Health:2
Flammability:1
Instability:0
DOTInformation DOT Class 6.1, : Poison