missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, Analytical Standard, ≥99.9% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018807
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018807 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018807 Supplier Merck Emd Millipore Supplier No. 024875ML
Chloroform is a colorless, volatile, liquid derivative of trichloromethane with an ether-like odor. It is a commonly used laboratory solvent. Presently, it is used in the industry as a solvent and in the production of the refrigerant freon. It is majorly used in production of chlorodifluoromethane, which is a refrigerant and fluoropolymer feedstock. It degrades in air to produce phosgene, dichloromethane, formyl chloride, CO, CO2 and hydrogen chloride. It can be orally consumed or inhaled, as it can be quickly absorbed, metabolized and eliminated by mammals.

Specifications

CAS 67-66-3
Solution Contains ˜0.003% amylene as stabilizer
Vapor Pressure 160 mmHg (20°C)
Vapor Density 4.1 (vs. air)
Density 1.492 g/mL (at 25°C)
Linear Formula CHCl3
MDL Number MFCD00000826
Melting Point -63°C
Boiling Point 60.5°C to 61.5°C
Synonym Methylidyne trichloride; Trichloromethane
Quantity 5 mL
Formula Weight 119.38
Percent Purity ≥99.9% (GC)
Grade Analytical Standard
Show More Show Less