missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, ACS Reagent, for HPLC, Spectrophotometry, meets analytical spec. of USP, contains amylene, >99.9%, Honeywell™

Catalog No. p-7112002
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
Poly-coated Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6000407 4 x 4 L Glass Bottle
6000408 4 x 4 L Poly-coated Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6000407 Supplier Honeywell Chemicals Supplier No. AH0494
Only null left

Chemical Identifiers

CAS 67-66-3 , 513-35-9
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

CAS Max % 99.9900%, <0.0100%
Density 1.49 g/cm3
Boiling Point 62°C
Quantity 4 x 4 L
Packaging Glass Bottle
UN Number UN1888
Beilstein 1731042
Water 0.02% max.
Solubility Information 81.5% in Water at 20°C
Formula Weight 119.37
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Chloroform, ACS Reagent, for HPLC, Spectrophotometry, meets analytical specification of USP, contains amylene, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Chloroform