missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, ACS Grade, ≥ 99.8%, LabChem™

Catalog No. p-5379504
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC130401 500 mL Amber Glass
LC130402 1 L Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC130401 Supplier LabChem Supplier No. LC130401
Only null left

Chemical Identifiers

CAS 67-66-3 , 64-17-5
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name trichloromethane
SMILES ClC(Cl)Cl

Specifications

Melting Point -64°C
Color Colorless
CAS Max % 99, 1
Decomposition Information Chlorine
Density 1,490kg/m3
Quantity 500 mL
Identification Passes Test
Packaging Amber Glass
Physical Form Liquid
Residue after Evaporation ≤0.001%
Specific Gravity 1490kg/m3
Linear Formula CHCl3
UN Number UN1888
Boiling Point 61°C
Formula Weight 119.38
Percent Purity ≥99.8%
Grade ACS
Chemical Name or Material Chloroform
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic if inhaled.
Causes skin and serious eye irritation.
Suspected of causing cancer.
Suspected of damaging the unborn child.
Causes damage to organs (liver, kidneys) through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe vapors.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If swallowed: Rinse mouth.
Call a poison center or doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation occurs: Get medical attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center or doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 200-663-8
Recommended Storage Room Temperature