missing translation for 'onlineSavingsMsg'
Learn More

Chlorodiphenylphosphine 97.0+%, TCI America™

Catalog No. C059725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C059725G 25 g
C0597100G 100 g
C0597500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C059725G Supplier TCI America Supplier No. C059725G

Chemical Identifiers

CAS 1079-66-9
Molecular Formula C12H10ClP
Molecular Weight (g/mol) 220.64
MDL Number MFCD00000529
InChI Key XGRJZXREYAXTGV-UHFFFAOYSA-N
Synonym chlorodiphenylphosphine, diphenylchlorophosphine, diphenylphosphinous chloride, phosphinous chloride, diphenyl, p-chlorodiphenylphosphine, diphenylphosphine chloride, phosphine, chlorodiphenyl, unii-wo975pjk1y, pph2cl, chloro diphenyl phosphane
PubChem CID 66180
IUPAC Name chlorodiphenylphosphane
SMILES ClP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 215°C
Quantity 25 g
UN Number 3265
Formula Weight 220.64
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Chlorodiphenylphosphine
EINECSNumber (3)-3943
TSCA Yes