Learn More
Chlorodiphenylphosphine, 98+%
CAS: 1079-66-9 | C12H10ClP | 220.64 g/mol
Supplier: Thermo Scientific Chemicals 04135218
Description
Chlorodiphenylphosphine is used in the synthesis of various phosphines and an useful reagent in for the introduction of Ph2P group. It is also useful as an intermediate to make antioxidants, flame retardants, stabilizers, catalysts, photoinitiators and optical brighteners. It is also used to produce diphenylphosphinous acid methyl esters. It is used as a precursor in the preparation of various phosphines. It is involved in the preparation of phosphines, which is used as stabilizers for plastics, flame retardants and UV-hardening paint systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Chlorodiphenylphosphine | |
| 14°C to 16°C | |
| 1.204 g/mL | |
| 138°C (280°F) | |
| C12H10ClP | |
| (C6H5)2PCl | |
| 50 g | |
| 512032 | |
| chlorodiphenylphosphine, diphenylchlorophosphine, diphenylphosphinous chloride, phosphinous chloride, diphenyl, p-chlorodiphenylphosphine, diphenylphosphine chloride, phosphine, chlorodiphenyl, unii-wo975pjk1y, pph2cl, chloro diphenyl phosphane | |
| XGRJZXREYAXTGV-UHFFFAOYSA-N | |
| chloro(diphenyl)phosphane | |
| 66180 | |
| ≥98% |
| 1079-66-9 | |
| Colorless to Yellow | |
| 318°C to 321°C | |
| Acrid | |
| 1.636 | |
| MFCD00000529 | |
| UN3265 | |
| Air and moisture sensitive | |
| Miscible with alcohol. Slightly miscible with ammonia. | |
| ClP(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 220.64 | |
| 220.64 | |
| Liquid |
Chemical Identifiers
| 1079-66-9 | |
| 220.64 | |
| XGRJZXREYAXTGV-UHFFFAOYSA-N | |
| 66180 | |
| ClP(C1=CC=CC=C1)C1=CC=CC=C1 |
| C12H10ClP | |
| MFCD00000529 | |
| chlorodiphenylphosphine, diphenylchlorophosphine, diphenylphosphinous chloride, phosphinous chloride, diphenyl, p-chlorodiphenylphosphine, diphenylphosphine chloride, phosphine, chlorodiphenyl, unii-wo975pjk1y, pph2cl, chloro diphenyl phosphane | |
| chloro(diphenyl)phosphane |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
H290-H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 214-093-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only