missing translation for 'onlineSavingsMsg'
Learn More

Chlorodiphenylphosphine, 98+%

Catalog No. AA4135209
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4135209 10 g
AA4135218 50 g
AA4135230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4135209 Supplier Thermo Scientific Chemicals Supplier No. 04135209

CAS: 1079-66-9 | C12H10ClP | 220.64 g/mol

Chlorodiphenylphosphine is used in the synthesis of various phosphines and an useful reagent in for the introduction of Ph2P group. It is also useful as an intermediate to make antioxidants, flame retardants, stabilizers, catalysts, photoinitiators and optical brighteners. It is also used to produce diphenylphosphinous acid methyl esters. It is used as a precursor in the preparation of various phosphines. It is involved in the preparation of phosphines, which is used as stabilizers for plastics, flame retardants and UV-hardening paint systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1079-66-9
Molecular Formula C12H10ClP
Molecular Weight (g/mol) 220.64
MDL Number MFCD00000529
InChI Key XGRJZXREYAXTGV-UHFFFAOYSA-N
Synonym chlorodiphenylphosphine, diphenylchlorophosphine, diphenylphosphinous chloride, phosphinous chloride, diphenyl, p-chlorodiphenylphosphine, diphenylphosphine chloride, phosphine, chlorodiphenyl, unii-wo975pjk1y, pph2cl, chloro diphenyl phosphane
PubChem CID 66180
IUPAC Name chloro(diphenyl)phosphane
SMILES ClP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 14°C to 16°C
Color Colorless to Yellow
Density 1.204 g/mL
Boiling Point 318°C to 321°C
Flash Point 138°C (280°F)
Odor Acrid
Refractive Index 1.636
Linear Formula (C6H5)2PCl
Quantity 10 g
UN Number UN3265
Beilstein 512032
Sensitivity Air and moisture sensitive
Solubility Information Miscible with alcohol. Slightly miscible with ammonia.
Formula Weight 220.64
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Chlorodiphenylphosphine
Show More Show Less
Hazard Category H290-H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 214-093-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only