missing translation for 'onlineSavingsMsg'
Learn More

Chlorodiphenylphosphine, 95%, tech.

Catalog No. AC109130250
Change view
Click to view available options
Quantity:
25 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC109130250 25 mL Glass bottle
AC109135000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC109130250 Supplier Thermo Scientific Chemicals Supplier No. 109130250

CAS: 1079-66-9 | C12H10ClP | 220.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent

Chemical Identifiers

CAS 1079-66-9
Molecular Formula C12H10ClP
Molecular Weight (g/mol) 220.64
MDL Number MFCD00000529
InChI Key XGRJZXREYAXTGV-UHFFFAOYSA-N
Synonym chlorodiphenylphosphine, diphenylchlorophosphine, diphenylphosphinous chloride, phosphinous chloride, diphenyl, p-chlorodiphenylphosphine, diphenylphosphine chloride, phosphine, chlorodiphenyl, unii-wo975pjk1y, pph2cl, chloro diphenyl phosphane
PubChem CID 66180
SMILES ClP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Density 1.2290g/mL
Boiling Point 320.0°C
Flash Point 138°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.6340 to 1.6370
Linear Formula (C6H5)2PCl
Quantity 25 mL
Beilstein 16, 763
Fieser 11,120; 16,77
Specific Gravity 1.229
Merck Index 15, 2139
Solubility Information Solubility in water: reacts violently with water
IUPAC Name chlorodiphenylphosphane
Formula Weight 220.64
Percent Purity 95%
Grade Technical
Chemical Name or Material Chlorodiphenylphosphine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
Reacts violently with water.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 214-093-2

RUO – Research Use Only