missing translation for 'onlineSavingsMsg'
Learn More

chlorodiphenylmethane, 98%

Catalog No. ac154662500
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mL
5 mL
50 mL
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC154662500 250 mL Glass bottle
AC154660050 5 mL Glass bottle
AC154660500 50 mL Glass bottle
3 options

Catalog No. AC154662500

Supplier: Thermo Scientific Chemicals 154662500

Only null left

CAS: 90-99-3 | C13H11Cl | 202.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 90-99-3
Molecular Formula C13H11Cl
Molecular Weight (g/mol) 202.68
MDL Number MFCD00000855
InChI Key ZDVDCDLBOLSVGM-UHFFFAOYSA-N
Synonym benzhydryl chloride, chlorodiphenylmethane, chloromethylene dibenzene, diphenylchloromethane, diphenylmethyl chloride, benzene, 1,1'-chloromethylene bis, chloro phenyl methyl benzene, methane, chlorodiphenyl, unii-cn9n9ayv4b, 1,1'-chloromethylene bisbenzene
PubChem CID 7035
IUPAC Name [chloro(phenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)Cl

Specifications

Melting Point 15°C to 17°C
Color Colorless to Yellow
Density 1.1400g/mL
Boiling Point 270°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5940 to 1.597
Linear Formula (C6H5)2CHCl
Quantity 250 mL
Specific Gravity 1.14
Solubility Information Solubility in water: slightly soluble
Formula Weight 202.68
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Chlorodiphenylmethane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 202-031-7

RUO – Research Use Only