missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethylvinylsilane 97.0+%, TCI America™

Catalog No. p-7124855
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C120825ML 25 mL
C1208100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C120825ML Supplier TCI America Supplier No. C120825ML
Only null left

Chemical Identifiers

CAS 1719-58-0
Molecular Formula C4H8ClSi
Molecular Weight (g/mol) 119.64
MDL Number MFCD00018090
InChI Key RABBDIBWJVOAPB-ONEGZZNKSA-N
Synonym chlorodimethylvinylsilane, dimethylvinylchlorosilane, vinyldimethylchlorosilane, chloro dimethyl vinylsilane, silane, chloroethenyldimethyl, chlorodimethyl vinyl silane, unii-xja6n5221t, chloroethenyldimethyl-silane, chloro ethenyl dimethylsilane, chloro-dimethyl-ethenyl-silane
PubChem CID 519368
IUPAC Name [(1E)-2-chloroethenyl]dimethylsilyl
SMILES C[Si](C)\C=C\Cl

Specifications

Boiling Point 82°C
Quantity 25 mL
UN Number 2985
Formula Weight 120.65
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Chlorodimethylvinylsilane
EINECSNumber (2)-2042
TSCA Yes