accessibility menu, dialog, popup

UserName

CAS RN 1719-58-0

IUPAC Name: [(1E)-2-chloroethenyl]dimethylsilyl
Synonyms: [(1E)-2-chloroethenyl]dimethylsilyl
Molecular Weight (g/mol): 119.64
Molecular Formula: C4H8ClSi
InChi Key: RABBDIBWJVOAPB-ONEGZZNKSA-N
SMILES: C[Si](C)\C=C\Cl
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)