missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethylvinylsilane, 97%

Catalog No. AAL1619814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1619814 25 g
AAL1619822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1619814 Supplier Thermo Scientific Chemicals Supplier No. L1619814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1719-58-0 | C4H8ClSi | 119.64 g/mol

Chlorodimethylvinylsilane is used to prepare silicon-containing polymers, silaheterocycles and new chelating ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorodimethylvinylsilane is used to prepare silicon-containing polymers, silaheterocycles and new chelating ligands.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to moisture. Store under dry inert gas.

Chemical Identifiers

CAS 1719-58-0
Molecular Formula C4H8ClSi
Molecular Weight (g/mol) 119.64
MDL Number MFCD00018090
InChI Key RABBDIBWJVOAPB-ONEGZZNKSA-N
Synonym chlorodimethylvinylsilane, dimethylvinylchlorosilane, vinyldimethylchlorosilane, chloro dimethyl vinylsilane, silane, chloroethenyldimethyl, chlorodimethyl vinyl silane, unii-xja6n5221t, chloroethenyldimethyl-silane, chloro ethenyl dimethylsilane, chloro-dimethyl-ethenyl-silane
PubChem CID 519368
SMILES C[Si](C)\C=C\Cl

Specifications

Boiling Point 82°C to 83°C
Quantity 25 g
UN Number UN2985
Beilstein 1737688
Flash Point −12°C (10°F)
Solubility Information Reacts with water.
IUPAC Name chloro-ethenyl-dimethylsilane
Formula Weight 120.66
Percent Purity 97%
Refractive Index 1.416
Sensitivity Moisture sensitive
Density 0.883
Chemical Name or Material Chlorodimethylvinylsilane
Show More Show Less
Hazard Category H225-H314
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P374-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROSILANES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 217-007-1
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only