missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethylsilane 95.0+%, TCI America™

Catalog No. p-7124604
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C077825ML 25 mL
C0778250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C077825ML Supplier TCI America Supplier No. C077825ML
Only null left

Chemical Identifiers

CAS 1066-35-9
Molecular Formula C2H6ClSi
Molecular Weight (g/mol) 93.61
MDL Number MFCD00000495
InChI Key QABCGOSYZHCPGN-UHFFFAOYSA-N
Synonym chlorodimethylsilane, dimethylchlorosilane, dmcs, silane, chlorodimethyl, unii-xr7e4f5838, chloro dimethyl silicon, chlorodimethyl-silan, dimethylmono-chlorosilane, chlorodimethylsilyl radical
PubChem CID 6327132
IUPAC Name chlorodimethylsilyl
SMILES C[Si](C)Cl

Specifications

Melting Point -111°C
Color Yellow
Boiling Point 37°C
Quantity 25 mL
UN Number 2985
Formula Weight 94.61
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Chlorodimethylsilane
EINECSNumber (2)-2041
TSCA Yes
Recommended Storage Refrigerator