accessibility menu, dialog, popup

UserName

CH3SiCH3Cl

CAS RN 1066-35-9

IUPAC Name: chlorodimethylsilyl
Synonyms: chlorodimethylsilane
Molecular Weight (g/mol): 93.61
Molecular Formula: C2H6ClSi
InChi Key: QABCGOSYZHCPGN-UHFFFAOYSA-N
SMILES: C[Si](C)Cl
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (5)