Learn More
Chlorodimethylsilane, 97%
CAS: 1066-35-9 | C2H6ClSi | 93.61 g/mol
Supplier: Thermo Scientific Chemicals A13113AK
Description
Chlorodimethylsilane is used to prepare vinyldimethylsilanols for palladium-catalyzed cross-coupling with aryl iodides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChlorodimethylsilane is used to prepare vinyldimethylsilanols for palladium-catalyzed cross-coupling with aryl iodides.
Solubility
Miscible with organic solvents.
Notes
It is incompatible with strong oxidizing agents, strong acids, strong bases, metals and water.
Specifications
| 34°C to 36°C | |
| -111°C | |
| 250 mL | |
| (CH3)2SiHCl | |
| UN2924 | |
| −28°C (−18°F) | |
| Miscible with organic solvents. | |
| C[Si](C)Cl | |
| 93.61 | |
| 94.62 | |
| Acrid | |
| Moisture sensitive |
| Chlorodimethylsilane | |
| 1066-35-9 | |
| C2H6ClSi | |
| MFCD00000495 | |
| 1633434 | |
| chlorodimethylsilane, dimethylchlorosilane, dmcs, silane, chlorodimethyl, unii-xr7e4f5838, chloro dimethyl silicon, chlorodimethyl-silan, dimethylmono-chlorosilane, chlorodimethylsilyl radical | |
| QABCGOSYZHCPGN-UHFFFAOYSA-N | |
| chloro(dimethyl)silicon | |
| 6327132 | |
| 97% | |
| 1.383 | |
| 0.869 |
Chemical Identifiers
| 1066-35-9 | |
| 93.61 | |
| QABCGOSYZHCPGN-UHFFFAOYSA-N | |
| 6327132 | |
| C[Si](C)Cl |
| C2H6ClSi | |
| MFCD00000495 | |
| chlorodimethylsilane, dimethylchlorosilane, dmcs, silane, chlorodimethyl, unii-xr7e4f5838, chloro dimethyl silicon, chlorodimethyl-silan, dimethylmono-chlorosilane, chlorodimethylsilyl radical | |
| chloro(dimethyl)silicon |
Safety and Handling
GHS H Statement
H224-H314-H318
Extremely flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H224-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber : 213-912-0
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only