missing translation for 'onlineSavingsMsg'
Learn More

Chlorodimethylsilane, 97%

Catalog No. aaa13113ad
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mL
50 mL
Catalog No. Quantity
AAA13113AD 50 mL
AAA13113AK 250 mL
2 options

Catalog No. AAA13113AD

Supplier: Thermo Scientific Chemicals A13113AD

Only null left

CAS: 1066-35-9 | C2H6ClSi | 93.61 g/mol

Chlorodimethylsilane is used to prepare vinyldimethylsilanols for palladium-catalyzed cross-coupling with aryl iodides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorodimethylsilane is used to prepare vinyldimethylsilanols for palladium-catalyzed cross-coupling with aryl iodides.

Solubility
Miscible with organic solvents.

Notes
It is incompatible with strong oxidizing agents, strong acids, strong bases, metals and water.

Chemical Identifiers

CAS 1066-35-9
Molecular Formula C2H6ClSi
Molecular Weight (g/mol) 93.61
MDL Number MFCD00000495
InChI Key QABCGOSYZHCPGN-UHFFFAOYSA-N
Synonym chlorodimethylsilane, dimethylchlorosilane, dmcs, silane, chlorodimethyl, unii-xr7e4f5838, chloro dimethyl silicon, chlorodimethyl-silan, dimethylmono-chlorosilane, chlorodimethylsilyl radical
PubChem CID 6327132
IUPAC Name chloro(dimethyl)silicon
SMILES C[Si](C)Cl

Specifications

Boiling Point 34°C to 36°C
Melting Point -111°C
Quantity 50 mL
Linear Formula (CH3)2SiHCl
UN Number UN2924
Beilstein 1633434
Flash Point −28°C (−18°F)
Solubility Information Miscible with organic solvents.
Formula Weight 94.62
Percent Purity 97%
Odor Acrid
Refractive Index 1.383
Sensitivity Moisture sensitive
Density 0.869
Chemical Name or Material Chlorodimethylsilane
Show More Show Less
Hazard Category H224-H314-H335
Hazard Statement GHS H Statement
H224-H314-H318
Extremely flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber 213-912-0
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only