missing translation for 'onlineSavingsMsg'
Learn More

chlorodifluoroacetic acid sodium salt, 96%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 404591000

Quantity 100g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC404591000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1895-39-2
152.46
MRTAVLDNYYEJHK-UHFFFAOYSA-M
2734985
C(=O)(C(F)(F)Cl)[O-].[Na+]
C2ClF2NaO2
MFCD00064771
sodium chlorodifluoroacetate, sodium 2-chloro-2,2-difluoroacetate, chlorodifluoroacetic acid sodium salt, sodium chloro difluoro acetate, acetic acid, chlorodifluoro-, sodium salt, sodiumchlorodifluoroacetate, difluorochloroacetic acid sodium salt, sodium difluorochloroacetate, chlorodifluoroacetic acid sodium
sodium;2-chloro-2,2-difluoroacetate

Specifications

chlorodifluoroacetic acid sodium salt
Authentic
Glass bottle
ClF2CCO2Na
MFCD00064771
sodium chlorodifluoroacetate, sodium 2-chloro-2,2-difluoroacetate, chlorodifluoroacetic acid sodium salt, sodium chloro difluoro acetate, acetic acid, chlorodifluoro-, sodium salt, sodiumchlorodifluoroacetate, difluorochloroacetic acid sodium salt, sodium difluorochloroacetate, chlorodifluoroacetic acid sodium
C(=O)(C(F)(F)Cl)[O-].[Na+]
152.46
152.46
1895-39-2
96%
C2ClF2NaO2
100g
01,1227; 02,379; 03,265; 05,603
MRTAVLDNYYEJHK-UHFFFAOYSA-M
sodium;2-chloro-2,2-difluoroacetate
2734985
96%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 217-586-

SDS
Documents

Documents