missing translation for 'onlineSavingsMsg'
Learn More

chlorodifluoroacetic acid sodium salt, 96%, Thermo Scientific™

Numéro de catalogue. AC404590050
Modifier l'affichage
Click to view available options
Quantité:
5g
25g
100g
Emballage:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
AC404590050 5g Glass bottle
AC404590250 25g Glass bottle
AC404591000 100g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. AC404590050 Fournisseur Thermo Scientific Chemicals Code fournisseur 404590050

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Identifiants chimiques

CAS 1895-39-2
Formule moléculaire C2ClF2NaO2
Poids moléculaire (g/mol) 152.46
Numéro MDL MFCD00064771
Clé InChI MRTAVLDNYYEJHK-UHFFFAOYSA-M
Synonyme sodium chlorodifluoroacetate, sodium 2-chloro-2,2-difluoroacetate, chlorodifluoroacetic acid sodium salt, sodium chloro difluoro acetate, acetic acid, chlorodifluoro-, sodium salt, sodiumchlorodifluoroacetate, difluorochloroacetic acid sodium salt, sodium difluorochloroacetate, chlorodifluoroacetic acid sodium
PubChem CID 2734985
Nom de l’IUPAC sodium;2-chloro-2,2-difluoroacetate
SOURIRES C(=O)(C(F)(F)Cl)[O-].[Na+]

Spécifications

Nom ou substance chimique chlorodifluoroacetic acid sodium salt
Spectre infrarouge Authentic
Fourchette de pourcentages d’essai 96%
Emballage Glass bottle
Formule linéaire ClF2CCO2Na
Quantité 5g
Fieser 01,1227; 02,379; 03,265; 05,603
Poids de formule 152.46
Pourcentage de pureté 96%
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 217-586-

RUO – Research Use Only