missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorodiethylisopropylsilane 97.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 75°C |
| Quantity | 1 g |
| UN Number | 2986 |
| Formula Weight | 164.75 |
| Percent Purity | ≥97.0% (T) |
| Physical Form | Liquid |
| Chemical Name or Material | Chlorodiethylisopropylsilane |
Safety and Handling
| EINECSNumber | (2)-2041 |
| TSCA | No |