missing translation for 'onlineSavingsMsg'
Learn More

Chlorodiethylisopropylsilane 97.0+%, TCI America™

Catalog No. D22625G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22625G 5 g
D22621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22625G Supplier TCI America Supplier No. D22625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 107149-56-4
Molecular Formula C7H17ClSi
Molecular Weight (g/mol) 164.75
MDL Number MFCD00191631
InChI Key OSBPGFIPLLCQMO-UHFFFAOYSA-N
Synonym chlorodiethylisopropylsilane, diethylisopropylsilyl chloride, acmc-1bql9, isopropyldiethylchlorosilane, silane,chlorodiethyl 1-methylethyl, silane, chlorodiethyl 1-methylethyl
PubChem CID 4486980
IUPAC Name chlorodiethyl(propan-2-yl)silane
SMILES CC[Si](Cl)(CC)C(C)C

Specifications

Color Colorless
Boiling Point 75°C
Quantity 5 g
UN Number 2986
Formula Weight 164.75
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Chlorodiethylisopropylsilane
EINECSNumber (2)-2041
TSCA No