missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlorodiethylisopropylsilane, 95+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 435650250
| Quantity | 25mL |
|---|---|
| Packaging | Glass bottle |
Description
Chemical Identifiers
| 107149-56-4 | |
| 164.75 | |
| OSBPGFIPLLCQMO-UHFFFAOYSA-N | |
| 4486980 | |
| CC[Si](Cl)(CC)C(C)C |
| C7H17ClSi | |
| MFCD00191631 | |
| chlorodiethylisopropylsilane, diethylisopropylsilyl chloride, acmc-1bql9, isopropyldiethylchlorosilane, silane,chlorodiethyl 1-methylethyl, silane, chlorodiethyl 1-methylethyl | |
| chloro-diethyl-propan-2-ylsilane |
Specifications
| chlorodiethylisopropylsilane | |
| 0.9000g/mL | |
| 38°C | |
| 95% min. (GC) | |
| C7H17ClSi | |
| (CH3)2CHSi(C2H5)2Cl | |
| 25mL | |
| chlorodiethylisopropylsilane, diethylisopropylsilyl chloride, acmc-1bql9, isopropyldiethylchlorosilane, silane,chlorodiethyl 1-methylethyl, silane, chlorodiethyl 1-methylethyl | |
| OSBPGFIPLLCQMO-UHFFFAOYSA-N | |
| chloro-diethyl-propan-2-ylsilane | |
| 4486980 | |
| 95+% |
| 107149-56-4 | |
| 75.0°C | |
| Authentic | |
| Glass bottle | |
| 1.4370 to 1.4390 (20°C, 589nm) | |
| MFCD00191631 | |
| 0.9 | |
| Solubility in water: reacts | |
| CC[Si](Cl)(CC)C(C)C | |
| 164.75 | |
| 164.75 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Reacts violently with water.
Contact with water liberates toxic gas.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call
GHS Signal Word: Danger