missing translation for 'onlineSavingsMsg'
Learn More

Chlorodicyclohexylphosphine 97.0+%, TCI America™

Catalog No. p-7125110
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16581G 1 g
C16585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16581G Supplier TCI America Supplier No. C16581G
Only null left

Chemical Identifiers

CAS 16523-54-9
Molecular Formula C12H22ClP
Molecular Weight (g/mol) 232.73
MDL Number MFCD00014294
InChI Key AKJFBIZAEPTXIL-UHFFFAOYSA-N
Synonym chlorodicyclohexylphosphine, dicyclohexylchlorophosphine, dicyclohexylphosphinous chloride, dicyclohexylphosphine chloride, chlorodicyclohexyl phosphine, phosphinous chloride, dicyclohexyl, clpcy2, pcy2cl, chlorodicyclohexvlphosphine, dicyclohexylchlorophosphane
PubChem CID 2734135
IUPAC Name chlorodicyclohexylphosphane
SMILES ClP(C1CCCCC1)C1CCCCC1

Specifications

Color Yellow
Boiling Point 166°C
Quantity 1 g
UN Number 3265
Formula Weight 232.73
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Chlorodicyclohexylphosphine