missing translation for 'onlineSavingsMsg'
Learn More

Chlorocarbonylsulfenyl Chloride 97.0+%, TCI America™

Catalog No. C10435G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10435G 5 g
C104325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10435G Supplier TCI America Supplier No. C10435G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2757-23-5
Molecular Formula CCl2OS
Molecular Weight (g/mol) 130.97
MDL Number MFCD00000703
InChI Key MNOALXGAYUJNKX-UHFFFAOYSA-N
Synonym chlorocarbonylsulfenyl chloride, chlorothio formyl chloride, chloroformylsulfenyl chloride, chloro chlorosulfanyl methanone, chlorothioformyl chloride, chlorocarbonylsulphenyl chloride, chlorocarbonyl sulfenyl chloride, chlorocarbonyl sulphenyl chloride, carbonochloridothioic acid, anhydrosulfide with thiohypochlorous acid, methane, chloro chlorothio oxo
PubChem CID 75990
IUPAC Name chloro(chlorosulfanyl)methanone
SMILES ClSC(Cl)=O

Specifications

Color Yellow
Boiling Point 98°C
Quantity 5 g
UN Number 3265
Formula Weight 130.97
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Chlorocarbonylsulfenyl Chloride