accessibility menu, dialog, popup

UserName

ClSClO

CAS RN 2757-23-5

IUPAC Name: chloro(chlorosulfanyl)methanone
Synonyms: chloro(chlorosulfanyl)methanone
Molecular Weight (g/mol): 130.97
Molecular Formula: CCl2OS
InChi Key: MNOALXGAYUJNKX-UHFFFAOYSA-N
SMILES: ClSC(Cl)=O
Boiling Point 
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)